Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)

Catalog No. AAH2723803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2723803 1 g
AAH2723806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2723803 Supplier Thermo Scientific Chemicals Supplier No. H2723803
Add to Cart

CAS: 1272-25-9 | C15H15Tb | 354.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1272-25-9
Molecular Formula C15H15Tb
Molecular Weight (g/mol) 354.21
MDL Number MFCD03427139
InChI Key WFRJBVQIASPUDD-UHFFFAOYSA-N
Synonym tris cyclopentadienyl terbium, acmc-1bvr0, tris, c5-cyclopenta-2,4-dien-1-yl terbium
PubChem CID 20025873
IUPAC Name cyclopenta-1,3-diene;terbium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Tb+3]

Specifications

Melting Point 235°C
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 354.22
Percent Purity 99%
Assay (99.9%-Tb)
Chemical Name or Material Tris(cyclopentadienyl)terbium(III), (REO)
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber 215-051-6
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.