Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Diethylamino)Phosphine

Catalog No. 25318925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-318-925G 25 g
25-318-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 25-318-925G Supplier Sigma Aldrich Supplier No. 25318925G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(diethylamino)phosphine [(Et2N)3P] can be used as a reagent to synthesize: 1,1'sa-Dialkylisoindigo derivatives by reacting with various 1-alkylisatins via deoxygenation reaction.1-Aminomethylisatins by treating with isatin and primary and secondary amines.It can also be used in the deoxygenation of some cyclic +a-diketones using fullerene C60.

Specifications

CAS 2283-11-6
Density 0.903 g/mL at 25°C
Boiling Point 80°C to 90°C (10 mmHg, lit.)
Molecular Formula C12H30N3P
Refractive Index n20/D 1.475
Linear Formula P[N(C2H5)2]3
MDL Number MFCD00009041
Quantity 25 g
Synonym Hexaethyltriamidophosphite; Hexaethyltriaminophosphine; Phosphorous acid tris(diethylamide); Tris(N,N-diethylamino)phosphine; Tris(diethylamido)phosphine; Hexaethylphosphorous triamide
Molecular Weight (g/mol) 247.36 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.