Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dimethylamino)aluminum dimer

Catalog No. AA4323703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4323703 1 g
AA4323706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4323703 Supplier Thermo Scientific Chemicals Supplier No. 04323703
Add to Cart

Preparation of aluminum nitrides | CAS: 32093-39-3 | C12H36Al2N6 | 318.425 g/mol

Preparation of aluminum nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32093-39-3
Molecular Formula C12H36Al2N6
Molecular Weight (g/mol) 318.425
MDL Number MFCD00269811
InChI Key JGZUJELGSMSOID-UHFFFAOYSA-N
Synonym tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum
PubChem CID 16688802
IUPAC Name N-[bis(dimethylamino)alumanyl]-N-methylmethanamine
SMILES CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C

Specifications

Melting Point 87°C to 89°C
Color Brown to White
Density 0.865 g/mL
Boiling Point 90°C (0.05 mmHg)(sublimation)
Flash Point 21°C (70°F)
Linear Formula Al2[N(CH3)2]6
Quantity 1 g
UN Number UN3131
Sensitivity Moisture sensitive
Formula Weight 318.42
Physical Form Powder
Chemical Name or Material Tris (dimethylamino)aluminum dimer
Show More Show Less
Hazard Category H228-H260-H314-H335
Hazard Statement GHS H Statement
H228-H260-H314-H318
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
TSCA No
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.