Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dimethylamino)phosphine (may contain precipitate) 96.0+%, TCI America™
SDP

Catalog No. T1317250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1317250ML 250 mL
T131725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1317250ML Supplier TCI America Supplier No. T1317250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1608-26-0
Molecular Formula C6H18N3P
Molecular Weight (g/mol) 163.21
MDL Number MFCD00008301
InChI Key XVDBWWRIXBMVJV-UHFFFAOYSA-N
Synonym hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide
PubChem CID 15355
IUPAC Name [bis(dimethylamino)phosphanyl]dimethylamine
SMILES CN(C)P(N(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 163°C
Quantity 250 mL
UN Number 1993
Formula Weight 163.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(dimethylamino)phosphine (may contain precipitate)
RTECSNumber TH3390000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.