Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-HCl, Molecular Grade, Promega™

Catalog No. PRH5125 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRH5125 2500 g
PR-H5121 100 g
PR-H5123 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRH5125 Supplier Promega Corporation Supplier No. H5125
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in combination with Tris base for preparation of Tris-HCl buffers.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

For Use With (Application) Molecular Biology
pH 4.2 to 5.0 (0.1M)
Physical Form Crystalline
Quantity 2500 g
Solution Type Buffer
Formula Weight 157.56
Percent Purity ≥99.9%
Grade Molecular Biology
Chemical Name or Material Tris-HCL
Recommended Storage 15 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.