Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tris, Hydrochloride, ULTROL™ Grade, Calbiochem™,

Catalog No. 6483131KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-831-31KG 1 kg
64-831-3250GM 250 g
64-831-3500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 64-831-31KG Supplier MilliporeSigma™ Supplier No. 6483131KG
Add to Cart
Add to Cart

Useful in the pH of 3.5 to 5.0

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris(Hydroxymethyl)aminomethane, HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point 149°C
Quantity 1 kg
Solubility Information Soluble in water (800g/L at 20°C)
Formula Weight 157.6
Chemical Name or Material Tris, Hydrochloride
Grade by titration
Recommended Storage 15°C to 30°C (Do not freeze).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.