Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane Acetate 99.0+%, TCI America™
SDP

Catalog No. T3294100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3294100G 100 g
T329425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3294100G Supplier TCI America Supplier No. T3294100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6850-28-8
Molecular Formula C6H15NO5
Molecular Weight (g/mol) 181.188
MDL Number MFCD00038951
InChI Key PIEPQKCYPFFYMG-UHFFFAOYSA-N
Synonym tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PubChem CID 16218782
ChEBI CHEBI:66869
IUPAC Name acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES CC(=O)O.C(C(CO)(CO)N)O

Specifications

Melting Point 120°C
Color White
Quantity 100 g
Formula Weight 181.19
Percent Purity ≥99.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.