Learn More
Strem, An Ascensus Company CAS# 68959-87-5. 1g. Tris(i-propylcyclopentadienyl)lanthanum (99.9%-La) (REO). MFCD00145482
SDP
Supplier:  Strem, An Ascensus Company 5740001G
Encompass
CAS# 68959-87-5. 1g. Tris(i-propylcyclopentadienyl)lanthanum (99.9%-La) (REO). MFCD00145482. Molecular Weight: 460.43. Molecular Formula: (C3H7C5H4)3La. Color/form: colorless to pale yellow liq. Strem# 57-4000. http://www.strem.com/catalog/v/57-4000/
Catalog No. 50-204-5369