Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, MP Biomedicals™

Catalog No. MP21521761 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21521761 100 g
MP21521765 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21521761 Supplier MP Biomedicals, Inc Supplier No. 02152176.1

Used as a buffer in a wide variety of biological systems. Tris, MP Biomedicals provides pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis. It can be used as a starting material for polymers, oxazolones (w/ carbox

Tris is relatively non-hygroscopic; however, if needed, Tris (base) can be dried at 100°C for up to 4 hours to remove any water. Neither Tris nor Tris Hydrochloride by themselves provide adequate buffering capacity. Generally the two need to be mixed together to provide a buffer with pH between 7 and 9 to provide adequate buffering. The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per°C decrease in temperature, and decreases 0.03–0.05 unit per ten-fold dilution.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trometamol, Tromethamine, Tris-[hydroxymethyl]aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL)
Formula Weight 121.1
Percent Purity ≥99.0%
Grade Reagent
Recommended Storage Store at Room Temperature (15-30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.