Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS, MP Biomedicals™

Catalog No. MP219485590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219485590 500 g
MP219485580 100 g
MP219485583 250 g
MP219485591 1 kg
MP219485594 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP219485590 Supplier MP Biomedicals, Inc Supplier No. 0219485590
Add to Cart
Add to Cart

TRIS, Molecular Biology Reagent, ≥99.95%, White Crystalline Powder

  • Tris has been useful as buffers in a wide variety of biological systems.
  • Uses include pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
  • Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
  • Tris does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
  • It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
  • It is an auxiliary material in pharmaceutical science
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 167°C to 172°C
CAS 77-86-1
Boiling Point 219°C to 220°C at 10 mmHg (Literature)
Color White
pH 10 to 11.5 (5% aq. soln.)
Physical Form Crystalline Powder
Quantity 500 g
Purity Grade Notes ≥99.95%
Loss on Drying ≤1%
Molecular Formula C4H11NO3
MDL Number MFCD00004679
Synonym Trometamol, THAM, Tromethamine, Tris base
Solubility Information Soluble in water (40% aq soln) Clear, Colorless Solution (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL).
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Molecular Weight (g/mol) 121.14
Grade Molecular Biology Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.