Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(pentafluorophenyl)phosphine, 95%

Catalog No. AAL0274803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0274803 1 g
AAL0274806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0274803 Supplier Thermo Scientific Chemicals Supplier No. L0274803
Add to Cart

CAS: 1259-35-4 | C18F15P | 532.15 g/mol

Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Handle under inert gas. Incompatible with Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259-35-4
Molecular Formula C18F15P
Molecular Weight (g/mol) 532.15
MDL Number MFCD00079654
InChI Key FQLSDFNKTNBQLC-UHFFFAOYSA-N
Synonym tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane
PubChem CID 102076
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 115°C to 119°C
Odor Odorless
Quantity 1 g
Beilstein 773243
Solubility Information Insoluble in water.
Formula Weight 532.15
Percent Purity 97%
Chemical Name or Material Tris(pentafluorophenyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-021-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.