Learn More
Description
- Qualified for 2-D electrophoresis
- Tris (tris[hydroxymethyl] aminoethane)
- Prepares buffer with pH range of 7.2 to 9.0.
- Used in PAGE and Sequencing
- Soluble in cold water and hot water, methanol
- Solid physical form
Specifications
Specifications
| For Use With (Application) | Electrophoresis |
| pH | 7.2 to 9.0 |
| Physical Form | Solid |
| Quantity | 500 g |
| Solution Type | Buffer |
| Assay Percent Range | 99.8 to 100.1% |
| Formula Weight | 121.14 |
| Percent Purity | 99.8% |
| Grade | Electrophoresis |
| Chemical Name or Material | Tris, Suitable for preparing electrophoresis buffer in the range 7.2 to 9.0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.