Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyloxy)silane 98.0+%, TCI America™
SDP

Catalog No. T352025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T352025ML 25 mL
T35205ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T352025ML Supplier TCI America Supplier No. T352025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1873-89-8
Molecular Formula C9H27O3Si4
Molecular Weight (g/mol) 295.65
MDL Number MFCD00054865
InChI Key XAASNKQYFKTYTR-UHFFFAOYSA-N
Synonym tris trimethylsiloxy silane, tris trimethylsilyloxy silane, tris trimethylsilyloxy silicon, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, 1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, tris trimethylsilyloxy silyl radical, trisiloxane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy
PubChem CID 6365044
IUPAC Name 2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 mL
UN Number 1993
Formula Weight 296.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyloxy)silane
EINECSNumber (7)-0476&(7)-0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.