Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(triphenylphosphine)rhodium(I) chloride, 98%

Catalog No. p-209734
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190420010 1 g Glass bottle
AC190420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190420010 Supplier Thermo Scientific Chemicals Supplier No. 190420010
Only null left
Add to Cart

CAS: 14694-95-2 | C54H45ClP3Rh | 925.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Molecular Weight (g/mol) 925.23
MDL Number MFCD00010016
InChI Key IXAYKDDZKIZSPV-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
PubChem CID 84599
SMILES [Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 245.0°C to 250.0°C
Color Brown-Red to Maroon
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]3RhCl
Quantity 1 g
Fieser 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
Merck Index 15, 10245
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane
IUPAC Name λ¹-rhodium(1+) tris(triphenylphosphane) chloride
Formula Weight 925.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 238-744-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.