Learn More
Cayman Chemical THUNDREXreme PhosphoRK1 4800
SDP
Supplier:  Cayman Chemical 5002914800 WELL
Encompass_Preferred
An ALK5 inhibitor (IC50 0.0015 UM) selective for ALK over TGF-BRII (IC50 > 15 UM) and a panel of 213 additional kinases at 1 UM inhibits TGF-B-induced increases in the nuclear translocation of phosphorylated SMAD2/3 in Mv 1 Lu cells (IC50 0.35 UM) decreases tumor growth in an MC-38 murine colon carcinoma model of tumor rechallenge at 3.75 mg/kg per day in combination with an antibody against PD-1 induces hyperproliferation of chondrocytes in the femoral epiphysis thickening of the cardiac atrioventricular valve and inflammation in the aortic vasculature in rats at 10 mg/kg per day
Catalog No. 50-289-4261
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.