Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540616 Supplier Invitrogen™ Supplier No. PA540616
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PHDKRKVARS LQVFEETGIS HSEFLHRQHT EEGGGPLGGP LKFSNPCILW Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%.

TRIT1 is responsible for the modification of A37 to isopentenyl A37 of both cytosolic and mitochondrial tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Trit1
Gene Accession No. Q9H3H1
Gene Alias 2310075G14Rik; AI314189; AI314635; AV099619; GRO1; hGRO1; IPP transferase; IPPT; IPT; IPTase; isopentenyl-diphosphate:tRNA isopentenyltransferase; MOD5; Trit1; tRNA dimethylallyltransferase; tRNA dimethylallyltransferase, mitochondrial; tRNA isopentenylpyrophosphate transferase; tRNA isopentenyltransferase; tRNA isopentenyltransferase 1; tRNA isopentenyltransferase, mitochondrial
Gene Symbols Trit1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TRIT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.