Learn More
Boston Bioproducts Inc Triton X-100 Lysis Buffer - 500ml
Supplier:  Boston Bioproducts Inc BP117500ML
Encompass_Preferred
The Triton X-100 Lysis Buffer is a widely used lysis buffer in molecular biology and biochemistry applications. Composed of buffering copmponents as well as Triton X-100, a nonionic surfactant, this lysis buffer solubilizes lipids and proteins, effectively breaking open cellular compartments. The Triton X-100 Lysis buffer is known for its effectiveness in cell fractionation extraction protocols, due to its ability to solubilize the endoplasmic reticulum, plasma membrane, and nuclear envelope.
Catalog No. NC9348851
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.