Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton(R) X-405, 70% Solution in Water, Spectrum™ Chemical
SDP

Catalog No. 18614597 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-597 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-614-597 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR140500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9036-19-5

Specifications

CAS Max % 1
pH 7.0 to 9.0
Quantity 500 mL
Packaging Poly Bottle
Grade Reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For professional use only. Not for household use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.