Learn More
Thermo Scientific™ Triton™ X-100 Surfact-Amps™ Detergent Solution
Description
Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Features of Triton X-100 Surfact-Amps Detergent Solution:
- Triton X-100—a nonionic detergent for use in various protein methods
- Accurate—precise 10% detergent solution in ultrapure water
- Easy-to-use—solution is simple to dispense and dilute for use
- Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
- Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles
Specifications for Triton X-100 Surfact-Amps Detergent Solution:
- Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
- Concentration: 10.0±1.0%
- Oxidants: ≤1.0μeq/mL
- Carbonyls: ≤1.0μeq/mL
- Suspended Solids: Residue present must not exceed Residue Reference.
Properties of Triton X-100:
- Molecular Weight: 647 g
- Detergent Class: Nonionic polyoxyethylene surfactant
- Aggregation Number: 140
- Micelle Molecular Weight: 90,000 g
- Critical Micelle Concentration (CMC): 0.24 mM (0.0155% w/v)
- Cloud Point: 64°C
- Dialyzable: No
Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.
Related Products
- Surfact-Amps™ Detergent Sampler (Cat. No. 28340)
Specifications
Specifications
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. |
| Form | Liquid |
| Product Line | Surfact-Amps, Triton |
| Reagent Type | Detergent Solution |
| Quantity | 1 L |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.