Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton™ X-100, MP Biomedicals

Catalog No. mp04807426 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04807426 500 mL
MP04807427 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04807426 Supplier MP Biomedicals, Inc Supplier No. 04807426
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • The principal uses of Triton™ X-100 is in industrial and household detergent applications and in emulsifying agents
  • It is employed in almost every type of liquid, paste, and powdered cleaning compound, ranging from heavy-duty industrial products to gentle detergents for fine fabrics
  • It is also an important ingredient of primary emulsifier mixtures used in the manufacture of emulsion polymers, stabilizers in latex polymers, and emulsifiers for agricultural emulsion concentrates, and wettable powders

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless to Yellow
Physical Form Viscous Liquid
Quantity 500 mL
Linear Formula (C2H4O)x . C14H22O
Solubility Information Miscible in Water, Polar Organic Solvents (alcohols, glycols, ethers, ketones, etc.) & Aromatic Hydrocarbons; Soluble in Inorganic Salt Solutions & Aqueous Mineral Acids; Insoluble in Aliphatic Hydrocarbons.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight 625 avg
Chemical Name or Material TRITON™ X-100
Recommended Storage Store at Room Temperature(15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.