Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton X-100™, Reagent, Reagents

Catalog No. 01336902
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-902 1 L
01-336-901 250 mL
01-336-900 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-902 Supplier Reagents Holdings Llc Supplier No. C23428001A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triton X-100™, Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula (CH3)3CCH2C(CH3)2C6H4(OCH2CH2)x
Molecular Weight (g/mol) 250.1933
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Octoxynol, Octoxinol
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Specific Gravity 1.065
Chemical Name or Material Triton X-100™, Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.