Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton™ X-114, MP Biomedicals™

Catalog No. 0219397180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-193-97180 100 mL
02-193-97183 250 mL
ICN19397190 500 mL
02-193-97191 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-193-97180 Supplier MP Biomedicals, Inc Supplier No. 0219397180
Add to Cart
Add to Cart

Triton™ X-114 is a surfactant used in industrial and household detergent applications and in emulsifying agents.

  • Triton™ X-114 is used in biochemical applications to solubilize and separate proteins due to its low cloud point (23°C).
  • At temperatures above the cloud point, detergent solutions separate into aqueous and detergent-enriched phases.
  • This makes Triton™ X-114 particularly useful in separating lipophilic proteins from hydrophilic proteins.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9036-19-5
Synonym Octylphenoxypoly (ethoxyethanol), Alkylphenol hydroxypolyethylene, (1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol tert-octylphenyl ether, tert-octylphenoxypoly (ethoxyethanol)

Specifications

Color Light Yellow
Physical Form Liquid
Density 8.8 lb./gal
Quantity 100 mL
Packaging Plastic Container
Specific Gravity 1.054 g/ml
Molecular Formula (C2H4O)x · C14H22O (x= 7-8)
Molecular Weight (g/mol) 294.435 g/mol
Chemical Name or Material Triton™ X-114
Recommended Storage 15 to 30°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.