Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trityl Chloride Resin cross-linked with 1% DVB (200-400mesh) (2.0-2.5mmol/g), TCI America™
SDP

Catalog No. T34325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34325G Supplier TCI America Supplier No. T34325G

Chemical Identifiers

MDL Number MFCD00284810
Synonym Chlorodiphenylmethyl Polystyrene Resin

Specifications

Color Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Trityl Chloride Resin cross-linked with 1% DVB (200-400mesh) (2.0-2.5mmol/g)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.