Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tritylamine, 98%

Catalog No. AAA1386406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1386406 5 g
AAA1386403 1 g
AAA1386414 25 g
AAA1386422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1386406 Supplier Thermo Scientific Chemicals Supplier No. A1386406
Add to Cart

CAS: 5824-40-8 | C19H17N | 259.352 g/mol

Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

Solubility
Soluble in chloroform, methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5824-40-8
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
MDL Number MFCD00008047
InChI Key BZVJOYBTLHNRDW-UHFFFAOYSA-N
Synonym tritylamine, triphenylmethylamine, benzenemethanamine, .alpha.,.alpha.-diphenyl, tritylamin, trityl amine, aminotriphenylmethane, triphenylaminomethane, triphenyl methylamine, aminotrityl, polymer-bound
PubChem CID 138598
IUPAC Name triphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N

Specifications

Melting Point 100°C to 104°C
Boiling Point 223°C (14 mmHg)
Quantity 5 g
Beilstein 2113674
Solubility Information Soluble in chloroform,methanol. Insoluble in water.
Formula Weight 259.35
Percent Purity 98%
Chemical Name or Material Tritylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.