Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrkB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586622 Supplier Invitrogen™ Supplier No. PA586622
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses. TrkB is involved in the development and maintenance of the nervous system. Moreover, Trk B is a tyrosine kinase gene highly related to Trk A. Trk B expression is confined to tissues within the central and peripheral nervous systems. The brain-derived neurotrophic factor (BDNF) and NT-3, but not NGF, can induce rapid phosphorylation on tyrosine of Trk B gp145, one of the receptors encoded by Trk B, although BDNF elicits a response at least two orders of magnitude greater than NT-3. TrkB is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signaling through TrkB leads to cell differentiation. Mutations in the TrkB gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for the TrkB gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Ntrk2
Gene Accession No. P15209, Q16620, Q63604
Gene Alias AI848316; BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C030027L06Rik; EC 2.7.10.1; GP145 TrkB; GP145TrkB; GP145-TrkB; GP145-TrkB/GP95-TrkB; GP95 TrkB; GP95-TrkB; hyperphagia; kinase TrkB; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 2; neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; NTRK 2; Ntrk2; OTTHUMP00000021573; OTTHUMP00000021574; OTTHUMP00000021576; OTTHUMP00000021577; RATTRKB1; Tkrb; Trk B; TRKB; trk-B; trkB protein; trkB tyrosine kinase; TRKB1; Tropomyosin-related kinase B; tyrosine kinase receptor B
Gene Symbols Ntrk2
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 102-158 of Human Sarcoglycan-beta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18212, 25054, 4915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.