Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT2A Monoclonal Antibody (OTI3B7), TrueMAB™, OriGene
SDP

Catalog No. p-200012803
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9644 30 μL
50-167-5987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9644 Supplier OriGene Supplier No. TA505523S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRMT2A Monoclonal antibody specifically detects TRMT2A in Human samples. It is validated for Western Blot

Specifications

Antigen TRMT2A
Applications Western Blot
Classification Monoclonal
Clone OTI3B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Dixdc1
Gene Accession No. Q155Q3
Gene Alias HpaII tiny fragments locus 9c; hpaII tiny fragments locus 9c protein; Htf9c; Htf9-c; TRM2 tRNA methyltransferase 2 homolog A; TRM2 tRNA methyltransferase 2A; TRMT2A; tRNA (uracil-5-)-methyltransferase homolog A; tRNA methyltransferase 2 homolog A; tRNA methyltransferase 2 homolog A (S. cerevisiae)
Host Species Mouse
Immunogen Full length human recombinant protein of human DIXDC1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85458
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.