Learn More
TRMT2A Mouse anti-Human, Clone: OTI3B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505524

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by TRMT2A is of unknown function. However, it is orthologous to the mouse Trmt2a gene and contains an RNA methyltransferase domain. Expression of TRMT2A varies during the cell cycle, with aberrant expression being a possible biomarker in certain breast cancers. Several transcript variants encoding two different isoforms have been found for TRMT2A.Specifications
TRMT2A | |
Monoclonal | |
Unconjugated | |
TRMT2A | |
HTF9C | |
Mouse | |
Affinity Chromatography | |
RUO | |
27037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IZ69 | |
TRMT2A | |
Full length human recombit protein of human TRMT2A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.