Learn More
Research Products International Corp Transport Tube (17x60mm), 5mL, Attached WHT Screwcap, PP, Self-Standing, Printed Graduations, S, 20/bag, 25 bags/case
SDP
Supplier:  Research Products International Corp 163237SGL
Encompass
Ideal Transport Tubes are specially designed for sample collection, storage and transport of biological materials. Self-standing, transparent tubes are manufactured from shatter-proof, virgin polypropylene and withstand freezing temperatures and autoclaving. Feature external white threaded screw closure with a 2- 1/4 turn for fast opening or closing; clearly printed graduations; and white writing surfaces. Supplied with attached caps, 20 bags of 25
Catalog No. 50-222-9681