Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, UltraClass with NVLAP Certificate

Catalog No. 022142N Shop All Troemner Products
Change view
Click to view available options
Mass:
2 mg
3 mg
5 mg
30 mg
100 mg
200 mg
500 mg
1 g
3 g
5 g
10 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass
02-214-2N 100 mg
02-214-2A 100 g
02-214-2E 10 g
02-214-MW 200 g
02-214-2F 5 g
02-214-2G 3 g
02-214-2J 1 g
02-214-2K 500 mg
02-214-2M 200 mg
02-214-2Q 30 mg
02-214-2T 5 mg
02-214-2V 3 mg
02-214-2W 2 mg
02-214-116 5 kg
02-225-500 1 kg
02-225-534 500 g
02-225-535 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02-214-2N Supplier Troemner™ Supplier No. 30390245
Add to Cart
Add to Cart

Highly polished kilogram and gram weights

Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.

  • Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
  • Fractional weights 500mg to 50mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
  • The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
  • This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
  • Weights are supplied with a NIST/NVLAP Weight Calibration Certificate
  • Weights supplied in a rugged, precision-fit, high quality polycarbonate case
TRUSTED_SUSTAINABILITY

Specifications

Product Type One Piece Metric Analytical Sheet Weight
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass
Material Stainless Steel
Weight (Metric) 100 mg
Density 7.95g/cm3 at 20°C
Mass 100 mg
Shape Flat with one end turned up for easy handling
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.