Learn More
Enzo Life Sciences Troglitazone (5mg). CAS: 97322-87-7
SDP
Supplier:  Enzo Life Sciences BMLGR2100005
Encompass_Preferred
Antidiabetic thiazolidinedione. Potent and selective PPARγ ligand. Binds to the PPARγ ligand-binding domain and recruits ligand-specific transcriptional coactivators at 1µM. In skeletal muscle and adipocyte cell cultures, the EC50 of troglitazone is about 10µM. Displaces [125I]SB-236636 with an IC50=7.9µM. Stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells. Induces apoptosis in human vascular smooth muscle cells and inhibits the growth of a variety of human cancer cells including lung, bladder and myeloid leukemia cells. Displays an anti-invasive effect on human breast cancer cells; reduces migration, adhesion and spreading on fibronectin-coated plates. Inhibits cell growth of hematopoietic cell lines. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol (>25mg/ml, warm) or dimethyl formamide. Long Term Storage: Ambient.
Catalog No. 50-201-0932
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.