Learn More
Selleck Chemical LLC Troglitazone (CS-045) 10mg 97322-87-7 Rezulin, Romglizone, Prelay, CS-045, CS045, CS 045, Romozin
SDP
Supplier:  Selleck Chemical LLC S843210MG
Encompass_Preferred
Troglitazone (Rezulin, Romglizone, Prelay, CS045, Romozin) is a potent agonist for the peroxisome proliferator-activated receptor-(PPAR) that is a ligand activated transcription factor regulating cell differentiation and growth. Troglitazone induces autophagy, apoptosis and necroptosis in bladder cancer cells. Troglitazone prevents RSL3-induced ferroptosis and lipid peroxidation in Pfa1 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6055
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.