Learn More
Selleck Chemical LLC Trolox S3665-1g
SDP
Supplier:  Selleck Chemical LLC S36651G
Encompass_Preferred
Trolox is a cell-permeable water-soluble derivative of vitamin E with potent antioxidant properties
Catalog No. 50-310-8921
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.