Learn More
Chrono Log Corporation TROMBIN
SDP
Supplier:  Chrono Log Corporation 386
Encompass
This item has a minimum qty of 2 per supplier requirements.
Thrombin, Each vial contains a minimum of 10U of lyophilized thrombin from human plasma, Performs 10 to 20 tests
Catalog No. NC9864726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.