Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trop2 (EGP-1) Monoclonal Antibody (MR54), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 50245570
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-570 Supplier Invitrogen™ Supplier No. 14602482
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This MR54 monoclonal antibody reacts with human Trop-2, which is also known as Tumor-Associated Calcium Signal Transducer 2 (TACSTD2) and EGP-1. This type I transmembrane glycoprotein is expressed on trophoblast (syncytiotrophoblast and cytotrophoblast), epithelial, and most human carcinoma cells. Trop-2 plays a role in calcium-mediated signaling. Trop-2 has also been reported as an oncogene, with expression being correlated with tumorigenesis, metastasis, and anchorage-independent growth. Applications Reported: This MR54 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, western blotting, immunohistochemical staining of formalin-fixed paraffin embedded (FFPE) tissue sections (IHC-P), ELISA, and immunocytochemistry (ICC). Applications Tested: This MR54 antibody has been tested by flow cytometry and immunocytochemistry on MCF7 cells as well as by immunohistochemistry of formalin-fixed paraffin embedded tissue using either low or high pH antigen retrieval methods. Use of high pH antigen retrieval results in better staining intensity. This can be used at less than of equal to 5 μg/mL. Is is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trop2 (EGP-1)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MR54
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TACSTD2
Gene Accession No. P09758
Gene Alias 40kD glycoprotein, identified by monoclonal antibody GA733; C80403; cell surface glycoprotein TROP2; cell surface glycoprotein Trop-2; EGP1; EGP-1; epithelial glycoprotein-1; GA733; GA7331; GA733-1; gastrointestinal tumor-associated antigen GA7331; GP50; Ly97; lymphocyte antigen 97; M1S1; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; pancreatic carcinoma marker protein GA7331; pancreatic carcinoma marker protein GA733-1; parturition-related protein 1; Prp1; TACD2; TACSTD2; Trop2; truncated TACSTD2; tumor-associated calcium signal transducer 2; tunor-associated calcium signal transducer 2
Gene Symbols TACSTD2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4070
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.