Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TROP2 Monoclonal Antibody (A5E4)
GREENER_CHOICE

Catalog No. PIMA544974 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544974 Supplier Invitrogen™ Supplier No. MA544974
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

TROP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TROP2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A5E4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TACSTD2
Gene Accession No. P09758
Gene Alias 40kD glycoprotein, identified by monoclonal antibody GA733; C80403; cell surface glycoprotein TROP2; cell surface glycoprotein Trop-2; EGP1; EGP-1; epithelial glycoprotein-1; GA733; GA7331; GA733-1; gastrointestinal tumor-associated antigen GA7331; GP50; Ly97; lymphocyte antigen 97; M1S1; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; pancreatic carcinoma marker protein GA7331; pancreatic carcinoma marker protein GA733-1; parturition-related protein 1; Prp1; TACD2; TACSTD2; Trop2; truncated TACSTD2; tumor-associated calcium signal transducer 2; tunor-associated calcium signal transducer 2
Gene Symbols TACSTD2
Host Species Mouse
Immunogen Recombinant protein within human TROP2 aa 1-274/323 (Extracellular).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4070
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.