Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tropaeolin O, 0.04% w/v aq. soln.

Catalog No. AA42772AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42772AE 100 mL
AA42772AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42772AE Supplier Thermo Scientific Chemicals Supplier No. 042772AE
Add to Cart

CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym tropaeolin o
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 316.27
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Tropaeolin O
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.