Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropaeolin Oo, Solstice, Flauka™

Catalog No. 60045891 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-891 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-891 Supplier Solstice Supplier No. 310525G

Indicator, S. No.: 179

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-73-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.38
MDL Number MFCD00038130
InChI Key MLVYOYVMOZFHIU-UHFFFAOYSA-M
PubChem CID 23668839
IUPAC Name sodium;4-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Packaging Glass bottle
UN Number NONH for all modes of transport
Beilstein 3831566
Quantity 5 g
Physical Form Powder
Formula Weight 375.38g/mol
Chemical Name or Material Tropaeolin Oo, Indicator, S. No.: 179
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 209-071-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.