Learn More
Description
Tropicamide USP is a white or practically white crystalline powder. It is odorless or having not more than a slight odor.
Specifications
Specifications
| Chemical Name or Material | Tropicamide USP |
| Packaging | Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400 |
| CAS | 1508-75-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 284.357 |
| Quantity | 1 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
