Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropine 98.0+%, TCI America™
SDP

Catalog No. T05345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05345G 5 g
T053425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05345G Supplier TCI America Supplier No. T05345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-29-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00005551
InChI Key CYHOMWAPJJPNMW-RNFRBKRXSA-N
Synonym tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
PubChem CID 6101956
IUPAC Name (1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1C2CCC1CC(C2)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 141.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tropine
RTECSNumber YM3875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.