Learn More
Novus Biologicals™ Tropodithietic acid Broad spectrum antibiotic
Shop All Novus Biologicals Products

Description
Molecular Weight: 212.3
PubChem ID: 44632924
Application Notes: May be dissolved in DMSO (5 mg/mL).
Specifications
Specifications
| CAS | 750590-18-2 |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| For Use With (Application) | Orange Powder. May be Dissolved in DMSO (5 mg/mL). |
| Purity | >98% (HPLC); NMR (conforms) |
| Quantity | 1 mg |
| Source | Synthetic |
| Target | Tropodithietic acid Broad spectrum antibiotic |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.