Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropolone, 98%

Catalog No. AAA1129906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1129906 5 g
AAA1129903 1 g
AAA1129914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1129906 Supplier Thermo Scientific Chemicals Supplier No. A1129906
Add to Cart

Sensitive reagent for reducing sugars. A non-benzenoid aromatic compound, Reagent for the preparation of fused heterocycles and complexes of Ga(III) and In(III). | CAS: 533-75-5 | C7H6O2 | 122.123 g/mol

Tropolone is a sensitive reagent for reducing sugars. A non-benzenoid aromatic compound, Reagent for the preparation of fused heterocycles and complexes of Ga(III) and In(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00004158
InChI Key MDYOLVRUBBJPFM-UHFFFAOYSA-N
Synonym tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
PubChem CID 10789
ChEBI CHEBI:79966
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES C1=CC=C(C(=O)C=C1)O

Specifications

Melting Point 50°C to 55°C
Boiling Point 115°C (12 mmHg)
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 5 g
Beilstein 1904978
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tropolone
Show More Show Less
EINECSNumber 208-577-2
RTECSNumber GU4075000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.