Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Troponin I Recombinant Rabbit Monoclonal Antibody (1H11L19)

Catalog No. 701585
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-158-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-158-5 Supplier Invitrogen™ Supplier No. 701585
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. The TnI-fast and TnI-slow genes are expressed in fast-twitch and slow-twitch skeletal muscle fibers, respectively, while the TnI-cardiac gene is expressed exclusively in cardiac muscle tissue. This gene encodes the Troponin-I-skeletal-slow-twitch protein. This gene is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The encoded protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Troponin I
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H11L19
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene TNNI1
Gene Accession No. P13413, P19237, P19429, P23693, P48787, Q9WUZ5
Gene Alias 2700018B22Rik; AI747285; Cardiac troponin I; cardiomyopathy, dilated 2A (autosomal recessive); CMD1FF; CMD2A; CMH7; cTNI; DKFZp451O223; fast skeletal TnI; fsTnI; RCM1; slow skeletal TnI; slow skeletal troponin I isoform; ssTnI; Tn1; TnI; TNN1; TNNC1; TNNI1; TNNI3; Tpn1; troponin 1, type 3; troponin I type 1 (skeletal, slow); troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, skeletal, slow; troponin I, skeletal, slow 1; Troponin I, slow isoform; troponin I, slow skeletal; troponin I, slow skeletal muscle; Troponin I, slow-twitch isoform; troponin I1, slow skeletal type; troponin I3, cardiac type
Gene Symbols TNNI1, TNNI3
Host Species Rabbit
Immunogen human Troponin I.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21952, 21954, 29248, 29388, 7135, 7137
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.