Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TROY/TNFRSF19 Antibody Pair [HRP]
SDP

Catalog No. NB010111 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010111 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010111 Supplier Novus Biologicals™ Supplier No. NBP27953615PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 7.8 pg/mL
Assay Sensitivity 7.81-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse TROY/TNFRSF19.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Gene ID (Entrez) 55504
Gene Symbol TNFRSF19
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 130 ng of recombinant mouse TROY/TNFRSF19.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-mouse TROY/TNFRSF19 monoclonal antibody.
  • Detection Antibody: 1 mg/mL rabbit anti-TROY/TNFRSF19 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis
    Storage Requirements Storage is content dependent.
    Target TROY/TNFRSF19
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.