Learn More
CPC Scientific H-Thr-Arg-Pro-Ile-Ile-Thr-Thr-m-nitro-Tyr-Gly-Pro-Ser-Asp-Lys(Abz)-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HRV14002A
Encompass
This fluorogenic (FRET) substrate containing an anthraniloyl group and a 3-nitrotyrosine as the resonance energy transfer donor/quencher pair, was developed for the assay of 2A protease from human rhinovirus (kcat/Km = 154 M-1s-1), which has been viewed as an important enzyme target for antiviral intervention.ONE-LETTER SEQUENCE: TRPIITT-m-nitro-YGPSD-K(Abz)-YMOLECULAR FORMULA: C80H118N20O26MOLECULAR WEIGHT:1775.94STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [213135-91-2], RESEARCH AREA: AntimicrobialREFERENCES: Q.M.Wang et al., Arch. Biochem. Biophys., 356, 12 (1998)
Catalog No. 50-210-9577
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.