Learn More
Description
Immunogen sequence: IRRRTRKRLN PEALQAEQLN KQQRGSNEEQ VNGSPLERRS EDHLTESHQR EIPLPSLSKY EAQGSLTKSH SAQQPVLVSQ TLDIHKRM Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 98%.
Specifications
Specifications
| Antigen | TRPS1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRPS1 |
| Gene Accession No. | Q9UHF7 |
| Gene Alias | AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 |
| Gene Symbols | TRPS1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
