Learn More
TARGETMOL CHEMICALS INC TRPV4 AGONIST-1 FREE BAS 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T13214L
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TRPV4 agonist-1 free base (OUN67600) is an agonist of transient receptor potential vanilloid 4 (TRPV4;EC50 of 60 nM in the hTRPV4 Ca2+ assay). purity: 99%
Catalog No. 50-257-7402
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.