Learn More
Chemglass Life Sciences 10mm Tru-Stir™ Stirrer Shaft Coupling, 3/8" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck
SDP
Supplier:  Chemglass Life Sciences CG204401
Encompass_Preferred
10mm Tru-Stir™ Stirrer Shaft Coupling, 3/8" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck. The design of our Tru-Stir™ stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4” of offset misalignment and 45° of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10mm (OD) stirrer shafts. Upper adapters permit the coupling to be used with a standard Jacobs style 3-jaw chuck. Male Upper Coupling is 3/8". Overall length of the coupling is 4”. Supplied complete with an allen wrench set.
Catalog No. CG204401
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.