Learn More
Verichem Laboratories Tru-Zero Bilirubin Standard Kit 2 x 5mL
Supplier:  Verichem Laboratories 9456
Encompass_Preferred
Identical matrix as Verichem 9450 Bilirubin Standard Kit for optimum linearity, Protein based, ready-to-use liquid, No glycols, azides or surfactants, No special procedures or diluents required, Stable 5 days after opening when refrigerated
Catalog No. NC9941805
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.