Learn More
Perfex TruCLEAN 2
Catalog No. 19005382 Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Green
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-005-382 Yellow
19-005-379 -
19-005-380 -
19-005-381 Green
19-005-383 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-382 Supplier Perfex Supplier No. 302Y
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unique bucket-in-bucket design to simplify standard operating procedure and maximize operator adaptability.
Simplify standard operating procedures and maximize operator adaptability; all in a compact design for easy storage in confined areas.
Designed for small area cleaning and disinfecting, TruCLEAN 2’s bucket-in-bucket design isolates soiled contaminants from cleaning and disinfecting agents. Clean floors, walls, and ceilings with this truly efficient and lightweight mopping system. Temperature-resistant 36 Liter polypropylene bucket includes embossed graduations for accurate measure and 3-inch twin-wheel casters with one locking caster. Easily accessible brake pedal allows parking on uneven floors.
Specifications
Color Yellow