Learn More
Perfex TruCLEAN Pro Double-Bucket
Catalog No. 19005395 Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-005-395 White
19-005-394 Red
19-005-396 Blue
19-005-397 Green
19-005-398 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-395 Supplier Perfex Supplier No. 222W
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heavy-duty stainless-steel cart with multiple configurations, colorways, and accessories to customize for your specific environment.
Capture and isolate contaminants to ensure the delivery of unadulterated cleaning and sanitizing agents. No more re-applying dirty water and weakened solutions. TruCLEAN Pro systems efficiently clean and disinfect floors, walls, and ceilings in large or small area cleanrooms of any grade.
The adaptable TruCLEAN Pro system allows triple (15 L) or double (22 L) bucket configuration in a heavy-duty stainless-steel cart designed for strength, durability, and hygiene. Our Pro series offers the most versatile mopping equipment with multiple configurations, colorways, and accessories to customize for your specific environment.
Each mopping system component plays an important role in delivering the highest level of cleanliness. For example, our innovative swivel mop frame is accurately balanced to maintain total surface contact at any angle. The unique wringing mechanism exerts force evenly across the entire mop surface, extracting the maximum amount of soiled contaminants.
Specifications
Color White
Dimensions (L x W x H) 27 x 19 x 41 in.